Learn More
Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators
Description
Ensure accuracy and precision of your drug testing results by partnering these liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators with our Thermo Scientific DRI Hydrocodone Drugs of Abuse Assay and Control Set.These calibrators are for sale in the U.S.A. and Canada only.
- Specific detection of hydrocodone
- Qualitative or semi-quantitative modes
- For sale in the U.S.A. and Canada only
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | DRI Hydrocodone Drugs of Abuse Calibrator |
| Concentration | 500 ng/mL |
| Detectable Analytes | Hydrocodone |
| Sample Type | Urine |
| Type | Calibrator |
| Packaging | 1 Bottle |
| Quantity | 1 x 10 mL |
Disclaimers: Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.