Learn More
Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays
Description
Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers. Field-tested on a wide range of analyzers, these assays are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Control Sets | MGC Multi-Drug Controls |
| Description | DRI Methadone Drugs of Abuse Assays |
| Detectable Analytes | Methadone (MTD) |
| DoA Calibrators | Multi-drug Calibrator |
| Sample Type | Urine |
| Type | Reagent |
| Quantity | 1 x 500 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.