Learn More
Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators
Description
Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ Methadone Metabolite Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | DRI Methadone Metabolite Drugs of Abuse Calibrators |
| Concentration | 1000 ng/mL |
| Detectable Analytes | EDDP |
| Sample Type | Urine |
| Type | Calibrator |
| Packaging | 1 Bottle |
| Quantity | 1 x 10 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.