Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Calibrators

Catalog No. 23278403
Change view
Click to view available options
Quantity:
1 x 5 mL
1 x 25 mL
Concentration:
100 ng/mL
20 ng/mL
200 ng/mL
50 ng/mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
23-278-403 1 x 25 mL 50 ng/mL
23-278-302 1 x 5 mL 50 ng/mL
23-278-303 1 x 5 mL 100 ng/mL
23-278-310 1 x 5 mL 200 ng/mL
23-278-314 1 x 5 mL 20 ng/mL
23-278-404 1 x 25 mL 100 ng/mL
23-278-405 1 x 25 mL 200 ng/mL
23-278-402 1 x 25 mL 20 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 23-278-403 Supplier Thermo Scientific™ Supplier No. 1398
Add to Cart
Add to Cart
This item is not returnable. View return policy

Utilize the globally valued accuracy, precision and lot-to-lot dependability of DRI THC(Cannabinoid) Drugs of Abuse Calibrators.

Utilize the globally valued accuracy, precision and lot-to-lot dependability of Thermo Scientific™ DRI™ THC(Cannabinoid) Drugs of Abuse Calibrators, which are optimized for an array of clinical chemistry analyzers. The drugs of abuse testing scope is expanding rapidly, and DRI assays meet this challenge by offering one of the most extensive menus for drugs of abuse testing on the market.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI; 50ng/mL; 25mL
Concentration 50 ng/mL
Detectable Analytes 11-nor-9-COOH-D9-THC
Sample Type Urine
Type Calibrator
Packaging 1 Bottle
Quantity 1 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.