Learn More
Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls
Description
Validate the Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Assay and ensure accuracy of results with Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls. These liquid, ready-to-use controls streamline your testing process.
- One of the largest menus of tests available
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Applications for an array of clinical chemistry analyzers
- Variety of calibrators and multiconstituent control sets
Specifications
Specifications
| Description | DRI™ THC (Cannabinoid) Drugs of Abuse Control 75ng/mL, 5mL |
| Concentration | 75 ng/mL |
| Detectable Analytes | 11-nor-9-COOH-D9-THC |
| Sample Type | Urine |
| Type | Control |
| Quantity | 1 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.