Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls

Catalog No. 23278407
Change view
Click to view available options
Quantity:
1 x 5 mL
1 x 25 mL
Concentration:
125 ng/mL
40 ng/mL
60 ng/mL
75 ng/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
23-278-407 1 x 25 mL 60 ng/mL
23-278-307 1 x 5 mL 60 ng/mL
23-278-308 1 x 5 mL 40 ng/mL
23-278-311 1 x 5 mL 125 ng/mL
23-278-406 1 x 25 mL 40 ng/mL
23-278-409 1 x 25 mL 125 ng/mL
23-278-312 1 x 5 mL 75 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 23-278-407 Supplier Thermo Scientific™ Supplier No. 1402
Add to Cart
Add to Cart
This item is not returnable. View return policy

Eliminate time-consuming steps in reagent preparation with the Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls—ready-to-use controls in liquid form.

Validate the Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Assay and ensure accuracy of results with Thermo Scientific™ DRI™ THC (Cannabinoid) Drugs of Abuse Controls. These liquid, ready-to-use controls streamline your testing process.

  • One of the largest menus of tests available
  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Applications for an array of clinical chemistry analyzers
  • Variety of calibrators and multiconstituent control sets
TRUSTED_SUSTAINABILITY

Specifications

Description DRI™ THC (Cannabinoid) Drugs of Abuse Control 60ng/mL, 25mL
Concentration 60 ng/mL
Detectable Analytes 11-nor-9-COOH-D9-THC
Sample Type Urine
Type Control
Quantity 1 x 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.