Learn More
Andwin Scientific DRIERITE IND. 6 MESH 1 LB
SDP
Supplier:  Andwin Scientific 22890902
Encompass_Preferred
The Indicating DRIERITE Absorbent is an effective drying agent that conveniently provides visual indication of the progress of absorption and exhaustion. This product can be utilized at the end of large units of regular DRIERITE to signify the activity or exhaustion of regular DRIERITE. This can absorb 6-10% of its weight in water, contingent on relative humidity. As this product absorbs moisture, the indicator's color will change from blue to rose-red. This has a mesh size of 6 and a weight of 0.45 kg (1 lb).
Catalog No. NC9423911
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.