Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Flight Deck Jerseys
SDP

Catalog No. 17001116 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Color:
Blue
Brown
Green
Off White
Purple
Red
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
42 product options available for selection
Product selection table with 42 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-001-116 Green 2X-Large
17-001-100 Off White Small
17-001-101 Off White Medium
17-001-104 Off White 2X-Large
17-001-103 Off White X-Large
17-001-105 Off White 3X-Large
17-001-102 Off White Large
17-001-107 Yellow Medium
17-001-106 Yellow Small
17-001-108 Yellow Large
17-001-109 Yellow X-Large
17-001-113 Green Medium
17-001-111 Yellow 3X-Large
17-001-112 Green Small
17-001-110 Yellow 2X-Large
17-001-114 Green Large
17-001-115 Green X-Large
17-001-117 Green 3X-Large
17-001-118 Red Small
17-001-119 Red Medium
17-001-120 Red Large
17-001-122 Red 2X-Large
17-001-123 Red 3X-Large
17-001-124 Blue Small
17-001-121 Red X-Large
17-001-125 Blue Medium
17-001-126 Blue Large
17-001-127 Blue X-Large
17-001-128 Blue 2X-Large
17-001-131 Brown Medium
17-001-129 Blue 3X-Large
17-001-130 Brown Small
17-001-132 Brown Large
17-001-134 Brown 2X-Large
17-001-133 Brown X-Large
17-001-136 Purple Small
17-001-135 Brown 3X-Large
17-001-137 Purple Medium
17-001-139 Purple X-Large
17-001-140 Purple 2X-Large
17-001-138 Purple Large
17-001-141 Purple 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
42 options
This item is not returnable. View return policy
Catalog No. 17-001-116 Supplier DRIFIRE™ Supplier No. DF2246FDSGN2X
This item is not returnable. View return policy

Provide easy visual identification on shipboard flight decks

  • Lightweight DRIFIRE fabric dries four times faster that traditional cotton
  • Inherently flame-resistant — will not melt or fuse to skin when exposed to open flame
  • Mock turtleneck design increases coverage and protection
  • Thumbholes ensure sleeves stay in place
  • Excellent colorfastness — withstands standard shipboard laundry
  • Resists oil stains
  • Controls odor
  • Superior moisture wicking helps regulate skin temperature

Applications:

Industry: Navy.

Specifications

Size 2X-Large
Material Lightweight DRIFIRE fabric
Color Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.