Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Personal Protective Equipment
- Safety Clothing
- Shirts
- DRIFIRE™ Flight Deck Jerseys
DRIFIRE™ Flight Deck Jerseys
Catalog No. 17001120
Shop All National Public Seating Corporation Products

Change view
Click to view available options
Color:
Blue
Brown
Green
Off White
Purple
Red
Yellow
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
42 product options available for selection
| Catalog No. | Color | Size |
|---|---|---|
| 17-001-120 | Red | Large |
| 17-001-100 | Off White | Small |
| 17-001-101 | Off White | Medium |
| 17-001-104 | Off White | 2X-Large |
| 17-001-103 | Off White | X-Large |
| 17-001-105 | Off White | 3X-Large |
| 17-001-102 | Off White | Large |
| 17-001-107 | Yellow | Medium |
| 17-001-106 | Yellow | Small |
| 17-001-108 | Yellow | Large |
| 17-001-109 | Yellow | X-Large |
| 17-001-113 | Green | Medium |
| 17-001-111 | Yellow | 3X-Large |
| 17-001-112 | Green | Small |
| 17-001-110 | Yellow | 2X-Large |
| 17-001-116 | Green | 2X-Large |
| 17-001-114 | Green | Large |
| 17-001-115 | Green | X-Large |
| 17-001-117 | Green | 3X-Large |
| 17-001-118 | Red | Small |
| 17-001-119 | Red | Medium |
| 17-001-122 | Red | 2X-Large |
| 17-001-123 | Red | 3X-Large |
| 17-001-124 | Blue | Small |
| 17-001-121 | Red | X-Large |
| 17-001-125 | Blue | Medium |
| 17-001-126 | Blue | Large |
| 17-001-127 | Blue | X-Large |
| 17-001-128 | Blue | 2X-Large |
| 17-001-131 | Brown | Medium |
| 17-001-129 | Blue | 3X-Large |
| 17-001-130 | Brown | Small |
| 17-001-132 | Brown | Large |
| 17-001-134 | Brown | 2X-Large |
| 17-001-133 | Brown | X-Large |
| 17-001-136 | Purple | Small |
| 17-001-135 | Brown | 3X-Large |
| 17-001-137 | Purple | Medium |
| 17-001-139 | Purple | X-Large |
| 17-001-140 | Purple | 2X-Large |
| 17-001-138 | Purple | Large |
| 17-001-141 | Purple | 3X-Large |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 42 options available.
This item is not returnable.
View return policy
Catalog No.
17-001-120
Supplier
DRIFIRE™
Supplier No.
DF2246FDSRDLG
This item is not returnable.
View return policy
Description
- Lightweight DRIFIRE fabric dries four times faster that traditional cotton
- Inherently flame-resistant — will not melt or fuse to skin when exposed to open flame
- Mock turtleneck design increases coverage and protection
- Thumbholes ensure sleeves stay in place
- Excellent colorfastness — withstands standard shipboard laundry
- Resists oil stains
- Controls odor
- Superior moisture wicking helps regulate skin temperature
Applications:
Industry: Navy.
Specifications
Specifications
| Size | Large |
| Material | Lightweight DRIFIRE fabric |
| Color | Red |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?