Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Dungaree
SDP

Catalog No. 17010899 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size (English) Inseam:
30 in.
32 in.
Size (English) Waist:
32 in.
34 in.
36 in.
38 in.
40 in.
46 in.
50 in.
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English) Inseam Size (English) Waist
17-010-899 30 in. 32 in.
17-010-900 32 in. 32 in.
17-010-901 30 in. 34 in.
17-010-902 32 in. 36 in.
17-010-903 30 in. 38 in.
17-010-904 32 in. 38 in.
17-010-905 30 in. 40 in.
17-010-906 32 in. 40 in.
17-010-907 32 in. 46 in.
17-010-908 32 in. 50 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 17-010-899 Supplier National Safety Apparel™ Supplier No. PNTB2SD32X30
Add to Cart
Add to Cart

FR dungarees give a professional appearance while still remaining functional and offering lightweight comfort. Generous fit, large leg openings, and plenty of pockets.

  • Comfortable and generous fit in the seat and thighs for easy movement and all-day comfort
  • Ample storage with 8 pockets and 2 knife straps to keep tools and essentials within reach
  • Convenient phone storage with pockets designed to securely hold two large phones or other small devices
  • Relaxed fit design with wider leg openings to easily fit over work boots

Specifications

Gender Unisex
Product Type Flame-resistant Dungarees
Material FR Cotton
Color Navy
Seam Type Stitched
Closure Type Button, Zipper
Disposable Reusable
Hazard Risk Category CAT 2
Arc Flash Rating 10.7 cal/cm²
Certifications/Compliance NFPA 2112, NFPA 70E, OSHA 1910.269, ASTM F1506, CSA Z462
Size (English) Inseam 30 in.
Size (English) Waist 32 in.
Style Pants
For Use With (Application) Utilities, Oil, Gas
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.