Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Lightweight Long John
SDP

Catalog No. 17010858 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-858 X-Large
17-010-857 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-858 Supplier National Safety Apparel™ Supplier No. DF2446LPNBXL
Add to Cart
Add to Cart

FR long john base layer is designed for all day wear and features lightweight comfort, odor control, and breathability.

  • Lightweight, breathable fabric
  • Moisture wicking and quick dry rate
  • Inherently flame resistant, FR protection will not wash/wear out
  • Odor control technology
  • Soft fabric prevents chafing and maximizes comfort
  • FR Elastic waist maintains shape

Specifications

Size X-Large
Gender Unisex
Product Type Multirisk Pants
Material Modacrylic
Color Navy
Seam Type Stitched
No. of Pockets None
Disposable Reusable
Hazard Risk Category CAT 4
Arc Flash Rating 4.5 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, CSA Z462, OSHA 1910.269
Style Long John
For Use With (Application) Utilities, Electrical Contractors
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.