Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ FR Windbreaker Jacket
SDP

Catalog No. 17010861 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-861 Large
17-010-859 Small
17-010-860 Medium
17-010-862 X-Large
17-010-863 2X-Large
17-010-864 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-010-861 Supplier National Safety Apparel™ Supplier No. DF2CM1WWBNBLG
Add to Cart
Add to Cart

Jacket features wind, water, and cold resistance while providing breathability and protection.

  • Lightweight, breathable, and fast drying
  • Water and wind resistant
  • Resists cold cracking in low temperatures
  • Inset knitwrists
  • Jacket has zippered sleeve pocket and two lower hand warmer pockets
  • Made in the USA of globally sourced materials

Specifications

Size Large
Gender Unisex
Color Navy
Closure Type Zipper
Arc Flash Rating 37 cal/cm²
Certifications/Compliance OSHA 1910.269, ASTM F1959, ASTM F1506, CSA Z462, NFPA 70E
Disposable Reusable
Liner Material PYRAD™ Fabric by GORE-TEX LABS
Product Type FR Jacket
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.