Learn More
DRIFIRE™ Heavyweight Long Sleeve Shirts
SDP
Catalog No. 17000200 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Color:
Coyote Brown
Desert Sand
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-000-200 Desert Sand 3X-Large
17-000-189 Coyote Brown Small
17-000-192 Coyote Brown X-Large
17-000-191 Coyote Brown Large
17-000-190 Coyote Brown Medium
17-000-195 Desert Sand Small
17-000-194 Coyote Brown 3X-Large
17-000-193 Coyote Brown 2X-Large
17-000-196 Desert Sand Medium
17-000-199 Desert Sand 2X-Large
17-000-197 Desert Sand Large
17-000-198 Desert Sand X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 17-000-200 Supplier DRIFIRE™ Supplier No. DF2245LSDS3X
This item is not returnable. View return policy
Can be wear alone or as part of layering system for increased protection and warmth
  • Made of soft and comfortable, DRIFIRE fabric that prevents chafing
  • Suitable for cold climates
  • Flame resistant
  • Dropped-shoulder, flat-lock seams for no-rub comfort
  • Controls odor
  • Moisture-wicking and dries four times faster than cotton
Specifications
Size 3X-Large
Material Heavyweight; Flame resistant
Color Desert Sand