Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ HYDROflash FR Rain Hood
SDP

Catalog No. 17010871 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-871 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-010-871 Supplier National Safety Apparel™ Supplier No. HYDROFLAHHN
Add to Cart
Add to Cart

HYDROFlash FR™ (2-Layer GORE-TEX PYRAD™ fabric) is the lightest weight waterproof, breathable flash fire rated garment available.

  • Lightweight, breathable, and fast drying
  • 100% waterproof and windproof
  • Resists cold cracking in low temperatures
  • Hot liquid and steam resistant
  • Petroleum and oil-based mud resistant
  • Detachable, snap-on jacket hood is sold separately
  • Hook and loop adjustment for secure fit

Specifications

Product Type Fire Protectant Hood
Size One Size
Color Navy
Closure Type Hook and Loop
Disposable Reusable
Arc Flash Rating 35 cal/cm²
Certifications/Compliance ASTM F2733, ASTM F1891, CSA Z462, ASTM F903, OSHA 1910.269
Includes Rain Hood
For Use With (Application) Oil, Gas, Petrochemical
Resistance Flame
Hood Type Attachable
Style Hood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.