Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ HYDROflash™ FR Rain Jacket
SDP

Catalog No. 17010880 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-880 Large
17-010-878 Small
17-010-879 Medium
17-010-881 X-Large
17-010-882 2X-Large
17-010-883 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-010-880 Supplier National Safety Apparel™ Supplier No. HYDROFLASHJNLARGE
Add to Cart
Add to Cart

Featuring waterproof, windproof, and steam resistant technology, this jacket will withstand the cold and allow wearers to stay warm and dry to tackle the toughest jobs.

  • HYDROFlash FR™ (2-Layer GORE-TEX PYRAD™ Fabric) is the lightest weight waterproof, breathable Flash Fire rated garment available
  • Lightweight, breathable, and fast drying
  • 100% waterproof and windproof
  • Resists cold cracking in low temperatures
  • Hot liquid and steam resistant
  • Petroleum and oil-based mud resistant
  • FR 3M™ Scotchlite™ Reflective Comfort Trim
  • Hook and loop straps at cuffs

Specifications

Size Large
Gender Unisex
Color Navy
Closure Type Zipper
Arc Flash Rating 30 cal/cm²
Certifications/Compliance ASTM F2733, ASTM F1891, CSA Z462, ASTM F903, OSHA 1910.269
Disposable Reusable
Liner Material 2-Layer GORE-TEX PYRAD™ Fabric
Product Type FR Jacket
Style Rain Jacket
For Use With (Application) Oil, Gas, Petrochemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.