Learn More
Description
- Made of proprietary flame resistant material with 27-P design
- Durable, cool and breathable fabric that minimizes heat buildup
- With thigh-vents to cool femoral artery
- Moisture wicking and fast-drying
- High tensile and tear strengths
- Navy and USMC “Safe to Fly”
- Meets NAVAIR specifications
- Available in khaki color with different sizes ranging from Small to XXX-Large
US Navy, US Coast Guard, US Marines, US Law Enforcement/ Defense Contractors and MRO (Maintence Repair Operations)
Specifications
Specifications
| Size | Small |
| Material | Fabric |
| Color | Khaki |
| Certifications/Compliance | NAVAIR |
| For Use With (Application) | US navy, US coast guard, US marines, US law enforcement/ defense contractors and MRO |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
