Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Navair One-Piece Flight Suits
SDP

Catalog No. 17000335 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-000-335 Medium
17-000-268 Small
17-000-270 Large
17-000-272 2X-Large
17-000-337 X-Large
17-000-339 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-000-335 Supplier DRIFIRE™ Supplier No. PHX17129450TNMDR
This item is not returnable. View return policy

With adjustable side tabs for a perfect fit around the torso

  • Made of proprietary flame resistant material with 27-P design
  • Durable, cool and breathable fabric that minimizes heat buildup
  • With thigh-vents to cool femoral artery
  • Moisture wicking and fast-drying
  • High tensile and tear strengths
  • Navy and USMC “Safe to Fly”
  • Meets NAVAIR specifications
  • Available in khaki color with different sizes ranging from Small to XXX-Large

US Navy, US Coast Guard, US Marines, US Law Enforcement/ Defense Contractors and MRO (Maintence Repair Operations)

Specifications

Size Medium
Material Fabric
Color Khaki
Certifications/Compliance NAVAIR
For Use With (Application) US navy, US coast guard, US marines, US law enforcement/ defense contractors and MRO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.