Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ DRIFIRE™ Performance FR Jean 32 in.
SDP

Catalog No. 17355147 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
5X-Large
6X-Large
Catalog No. Size
17-355-147 X-Small
17-355-150 Small
17-355-153 Medium
17-355-156 Large
17-355-159 X-Large
17-355-162 2X-Large
17-355-165 3X-Large
17-355-168 5X-Large
17-355-171 6X-Large
9 options
This item is not returnable. View return policy

Catalog No. 17-355-147

Supplier: National Safety Apparel™ DF2CM618JNDN30X32

This item is not returnable. View return policy

National Safety Apparel™ DRIFIRE™ Performance FR Jeans are the ultimate heavy duty performance utility pant.

  • CAT 2
  • Fabric/Material: 14.75 oz. High Performance Denim
  • Dual certified to protect against flash fire and arc flash hazards
  • Ultimate heavy duty performance utility pant
  • Breathable fabric which increases moisture management
  • 5 pocket style and classic cut
  • Double tool pocket on the right side
  • Inherently flame resistant
  • FR protection will not wash/wear out
  • Quickly identify compliance with exterior labeling
  • Dries up to 37% faster than competitors
  • Leg openings made to fit over work boots
  • Comfort washed
  • Made in the USA

Specifications

Size X-Small
Material Denim
Color Blue
No. of Pockets 4
Arc Flash Rating 15 cal/cm2
Certifications/Compliance NFPA 70E for Arc Flash, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269
Size (English) Inseam 32 in.
Size (English) Waist 30 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.