Learn More
National Safety Apparel™ DRIFIRE™ Performance FR Jean 34 in.
Shop All National Public Seating Corporation Products

Description
- CAT 2
- Fabric/Material: 14.75 oz. High Performance Denim
- Dual certified to protect against flash fire and arc flash hazards
- Ultimate heavy duty performance utility pant
- Breathable fabric which increases moisture management
- 5 pocket style and classic cut
- Double tool pocket on the right side
- Inherently flame resistant
- FR protection will not wash/wear out
- Quickly identify compliance with exterior labeling
- Dries up to 37% faster than competitors
- Leg openings made to fit over work boots
- Comfort washed
- Made in the USA
Specifications
Specifications
| Size | Large |
| Material | Denim |
| Color | Blue |
| No. of Pockets | 4 |
| Arc Flash Rating | 15 cal/cm2 |
| Certifications/Compliance | NFPA 70E for Arc Flash, ASTM F1506 for Arc Flash, CSA Z462 for Arc Flash (Canada), OSHA 1910.269 |
| Size (English) Inseam | 34 in. |
| Size (English) Waist | 36 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.