Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Two-Piece Navy Flight Suits
SDP

Catalog No. 17000340 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Closure Type:
Zipper
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Size
17-000-340 Zipper X-Small
17-000-276 Zipper Medium
17-000-278 Zipper X-Large
17-000-279 Zipper 2X-Large
17-000-341 Zipper Small
17-000-343 Zipper Large
17-000-346 Zipper 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 17-000-340 Supplier DRIFIRE™ Supplier No. PHX27129JA450TNXSR
This item is not returnable. View return policy

Designed for Naval and Marine Aviators with end-user input

  • Permanent, inherent flame resistance that won't wash or wear out
  • Controls odor
  • Increased durability and FR performance
  • Unique design features to improve fit, increase range of motion and reduce snag hazards
  • Navy and USMC “Safe to Fly”
  • Meets NAVAIR specifications
  • Available in khaki color in different sizes ranging from X-Small to XXX-Large

Two-Piece Navy Flight Suit Jackets:

  • Angled shoulder pockets for easy access
  • Adjustable waist tabs for customized torso fit

US Navy, US Coast Guard, US Marines and MRO (Maintence Repair Operations)

Specifications

Size X-Small
Material Modacrylic, Cotton
Color Khaki
Closure Type Zipper
Resistance Flame
Certifications/Compliance NAVAIR
Description Modacrylic, Cotton, Para Aramid™, Anti-stat
No. of Pockets 4
Size (English) Chest >33 in.
For Use With (Application) Naval and marine aviators with end-user input
Length (English) 28.75 to 31.6 in.
Length (English) Sleeve 24.5 to 26.5 in.
Length (Metric) 73 to 80.2 cm
Length (Metric) Sleeve 62.2 to 67.3 cm
Height (English) 5.25 to 6.25 ft.
Height (Metric) 160 to 190.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.