Learn More
Description
- Permanent, inherent flame resistance that won't wash or wear out
- Controls odor
- Increased durability and FR performance
- Unique design features to improve fit, increase range of motion and reduce snag hazards
- Navy and USMC “Safe to Fly”
- Meets NAVAIR specifications
- Available in khaki color in different sizes ranging from X-Small to XXX-Large
Two-Piece Navy Flight Suit Jackets:
- Angled shoulder pockets for easy access
- Adjustable waist tabs for customized torso fit
US Navy, US Coast Guard, US Marines and MRO (Maintence Repair Operations)
Specifications
Specifications
| Size | 3X-Large |
| Material | Modacrylic, Cotton |
| Color | Khaki |
| Closure Type | Zipper |
| Resistance | Flame |
| Certifications/Compliance | NAVAIR |
| Description | Modacrylic, Cotton, Para Aramid™, Anti-stat |
| No. of Pockets | 4 |
| For Use With (Application) | Naval and marine aviators with end-user input |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
