Learn More
Invitrogen™ DRIL1 Polyclonal Antibody

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ARID3A.
Specifications
Specifications
| Antigen | DRIL1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | ARID3A |
| Gene Accession No. | Q99856 |
| Gene Alias | ARID domain-containing 3A; ARID domain-containing protein 3A; ARID3A; AT rich interactive domain 3A (Bright like); AT rich interactive domain 3A (BRIGHT- like) protein; AT rich interactive domain 3A (BRIGHT-like); AT-rich interaction domain 3A; AT-rich interactive domain-containing protein 3A; B-cell regulator of IgH transcription; BRIGHT; dead ringer homolog 1; dead ringer-like 1; dead ringer-like protein 1; Dri1; DRIL1; DRIL3; DRX; E2F-binding protein 1; E2FBP1 |
| Gene Symbols | ARID3A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.