Learn More
Selleck Chemical LLC Dronedarone HCl S2114-1g
SDP
Supplier:  Selleck Chemical LLC S21141G
Encompass_Preferred
Dronedarone HCl (SR33589) is a multichannel blocker targeting potassium channel sodium channel and calcium channel used as an antiarrhythmic drug for treatment of atrial fibrillation (AF)
Catalog No. 50-311-0013
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.