Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dronedarone Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D46891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46891G 1 g
D4689200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46891G Supplier TCI America Supplier No. D46891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141625-93-6
Molecular Formula C31H45ClN2O5S
Molecular Weight (g/mol) 593.22
MDL Number MFCD00914940
InChI Key DWKVCQXJYURSIQ-UHFFFAOYSA-N
Synonym N-[2-Butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-5-benzofuranyl]methanesulfonamide Hydrochloride
PubChem CID 219025
IUPAC Name N-[2-butyl-3-[4-[3-(dibutylamino)propoxy]benzoyl]-1-benzofuran-5-yl]methanesulfonamide;hydrochloride
SMILES CCCCC1=C(C2=C(O1)C=CC(=C2)NS(=O)(=O)C)C(=O)C3=CC=C(C=C3)OCCCN(CCCC)CCCC.Cl

Specifications

Melting Point 144°C
Color White
Quantity 1 g
Formula Weight 593.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Dronedarone Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.