Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Drop Forged G Clamp
SCI_ED

Catalog No. S24536 Shop All Eisco Products
Change view
Click to view available options
Length (Metric):
200 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S24536 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24536 Supplier Eisco™ Supplier No. CH0938F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For woodworking, fabrication, apparatus set-up and lessons on pressure and torque.

  • Threaded screw opens 200mm
  • Topped with swivel pad for better grip
  • Sliding T-bar handle allows easy tightening and untightening

Specifications

Type G-Clamp
Material Steel
Includes Sliding T-bar handle for easy tightening and untightening
For Use With (Application) Threaded screw opens 200mm
Description Topped with swivel pad
Length (Metric) 200 mm
Color Black
Jaw Type Swivel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.