Learn More
United States Biological Corporation DROP OUT BASE/GALACTUSE/100GR
Catalog No. NC0931986 Shop All United States Biological Corporation Products
Encompass_Preferred
Drop-out Mix Synthetic Minus Tryptophan w/o Yeast Nitrogen Base (DO, Dropout, Powder)
Catalog No. NC0931986 Supplier United States Biological Corporation Supplier No. D9530100G
Drop-out Mix Synthetic Minus Tryptophan w/o Yeast Nitrogen Base (DO, Dropout, Powder)