Learn More
United States Biological Corporation Drop-out Base With Galactose (DO, Dropout, Powder) 100g
Catalog No. NC3735049 Shop All United States Biological Corporation Products
Encompass_Preferred
Dissolve 26.7g in 800-900ml of ddH2O stirring gently until completely solubilized. Adjust pH of the medium to the desired level. Add additional water to bring the solution to1L. Filter-sterilize using a 0.22 micron membrane filter. Aliquot into sterile containers. Do not autoclave. Contains heat-labile compounds that can be damaged with autoclaving.
Catalog No. NC3735049 Supplier United States Biological Corporation Supplier No. D9502100G
Dissolve 26.7g in 800-900ml of ddH2O stirring gently until completely solubilized. Adjust pH of the medium to the desired level. Add additional water to bring the solution to1L. Filter-sterilize using a 0.22 micron membrane filter. Aliquot into sterile containers. Do not autoclave. Contains heat-labile compounds that can be damaged with autoclaving.