Learn More
United States Biological Corporation Drop-out Mix Synthetic, Minus Methionine and Uracil w/o Yeast Nitrogen Base (DO, Dropout, Powder) 100g
Catalog No. NC3826099 Shop All United States Biological Corporation Products
Encompass_Preferred
Drop-out Mix Complete (DOC) is used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with Methods in Yeast Genetics.
Catalog No. NC3826099 Supplier United States Biological Corporation Supplier No. D951702C100G
Drop-out Mix Complete (DOC) is used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with Methods in Yeast Genetics.