Learn More
Med Vet International Specimen Collection and Transport System ESwab™ Sterile, 220245, 50/Box
Supplier: Med Vet International 661468BX
Specimen collection and transport system has a flocked swab tip and liquid amies medium additive: It's for aerobic, anaerobic and fastidious bacteria collection Sample rapidly elutes from the flocked swab when placed in the transport medium testing Provides 1 mL of sample suspension for gram stain and multiple culture analyses Suitable platform for automated liquid specimen handling systems
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.