Learn More
Med Vet International Foam Dressing ComfortFoam™ Border 7 X 7 Inch With Border Waterproof Backing Silicone Adhesive Square Sterile, 43770, 5/Box
Supplier: Med Vet International 946496BX
The 7- x 7-inch DermaRite ComfortFoam border facilitates faster and proper healing of wounds A multilayer design optimizes wound protection, thermal insulation and absorbency The skin-friendly adhesive foam dressing sticks well and removes easily A low-profile border improves adhesion and flexibility for application in difficult areas
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.