Learn More
Drummond™ Digital Microdispensers
Description
Includes
Microdispenser 21-169-20D is supplied with 50 glass bores; 21-169-10A is supplied with 7 polypropylene bores; all other models supplied with package of 100 glass bores.
One-hand-operated positive displacement pipetter with digital reading of delivery volume to 0.01, 0.1, or 1.0μL. Five adjustable models cover volume range 0.01 to 1000μL.
- Adjustable to any volume within capacity rating
- Easy-to-set micrometer-type barrel for exact setting of volume to be pipetted
- Accuracy to ±1% at full volume
- Machined slot in barrel allows visual check for glass bore seating
- Color-coded calibration mark on bore lines up with plunger at full-capacity setting
- Borosilicate glass bores with stainless-steel plunger (PTFE tips on 25μL or larger); polypropylene bore on 1000μL size
- Use with all reagents (except hydrofluoric acid)
Order Info
Microdispenser 21-169-10A converts to a multichannel dispenser with the addition of a stainless-steel manifold.
Specifications
Specifications
| Accuracy | ±1% |
| Color | Red |
| Number of Channels | 1 |
| Type | Digital Microdispenser |
| Volume (Metric) | 10 μL |
| For Use With (Application) | Positive lock prevents accidental volume changes |
| Includes | 2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide |
| Increments | 0.01 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.