Learn More
Drummond™ Fixed Volume Microdispensers Replacement Parts Kits
Catalog No. 2117112
Change view
Click to view available options
Volume (Metric):
10 μL
100 μL
100 to 200 μL
25 μL
250 μL
5 μL
50 μL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-171-12 25 μL
21-171-16 250 μL
21-171-10 10 μL
21-171-13 50 μL
21-171-15 100 to 200 μL
21-171-5 5 μL
21-171-14 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 21-171-12 Supplier Drummond™ Supplier No. 3000325K
Add to Cart
Add to Cart
This item is not returnable. View return policy
Replacement Parts Kits for Microdispensers. For contents of each kit, see chart below.
Specifications
Volume (Metric) 25 μL
For Use With (Equipment) Drummond™ Fixed Volume Microdispenser
Includes 1 Plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide