Learn More
Drummond™ Fixed Volume Microdispensers Replacement Parts Kits
Description
Replacement Parts Kits for Microdispensers. For contents of each kit, see chart below.
Specifications
Specifications
| Volume (Metric) | 100 μL |
| For Use With (Equipment) | Drummond™ Fixed Volume Microdispenser |
| Includes | 1 Plunger, 2 O-rings, 1 collet, 1 coil spring, 3 PTFE tip, 1 spring guide |