Learn More
Drummond™ Portable Pipet-Aid™ XL Pipet Controller Accessory, EZ Grip Nosepieces
Description
- Choice of colors: Orange, blue, red, violet, yellow, green
- Autoclavable
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Yellow |
| For Use With (Equipment) | Pipet-Aid™ XP, XL, and Hood Mate™ |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.