Learn More
Drummond™ Dispensing Manifold
Catalog No. 2116910D
Change view
Click to view available options
Angle:
No
Yes
Number of Channels:
8
12
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Angle Number of Channels
21-169-10D No 8
21-169-11B Yes 12
21-169-11A No 12
21-169-10E Yes 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-169-10D Supplier Drummond™ Supplier No. 3000093
Add to Cart
Add to Cart
Eight-tip manifolds for use with the 21-169-10A Digital Microdispenser or any pipetter that accepts standard Luer-Lok™ connector
  • Stainless steel and PTFE construction
  • Use for both sample delivery and aspiration operations
  • Supplied complete with cleaning wire for easy cleaning
  • Positive Luer lock connection
Specifications
Angle No
Description 9mm (center to center); 11mm pin length
Material Stainless Steel, PTFE
Number of Channels 8
For Use With (Equipment) Drummond™ micropipetter
Includes Cleaning wire, Luer connector and end caps