Learn More
Description
Dry block heaters require either one or two modular blocks, sold separately.
- Ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
- Integral support rod holder with locking knob accepts optional external temperature probe kit
- Accept separate interchangeable modular blocks (sold separately), accommodating various tube sizes from 0.2mL microtubes to 50mL centrifuge tubes
- Each block has a thermometer well for measuring block temperature (blocks sold separately)
- Efficient heater features close tube-and-block contact with a low-density design for exceptional temperature uniformity
- Temperature range: ambient 5° to 120°C
- With microprocessor control, LED display, audible alarm, temperature calibration mode
Applications:
Denaturing proteins, DNA applications, ELISA and other immunoassay studies
Order Info
Heaters require an Ohaus modular heating block for operation (sold separately). See S96607 for 1.5mL microcentrifuge tubes, S96608 for titer plates, S96609 for 15mL centrifuge tubes, S96610 for 50mL centrifuge tubes, S96611 for 10mm tubes, S96612 for 15/1
Specifications
Specifications
| Holds | 2 Blocks |
| Material | Aluminum |
| Temperature Range (Metric) | Ambient +5°C to 150°C |
| Type | Dry Block Heater |
| Heat Up Time | 50 min. |
| Includes | 92 in. Detachable, 3-wire cord and plug |
| Temperature Uniformity | ±0.1°C |
| Length (English) | 15.4 in. |
| Length (Metric) | 39.1 cm |
| Height (English) | 3.5 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
