Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Dry-Sponge with Gloves BP237SPG, Sold by AquaPhoenix Scientific
SCI_ED

Catalog No. S26789 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Dimensions (L x W):
1.5 x 3 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S26789 1.5 x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S26789 Supplier 3M™ Supplier No. BP237SPG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biocide-free cellulose sponge includes two polyethylene gloves and sample bag

  • 1.5 x 3 in. dry sponge can be used with wide range buffers and enrichment media and in dry-only environments
  • Simplified packaging convenience, including sponge, sample bag and gloves
    • Minimizes waste, handling and preparation
  • 5 x 9 in. sample bag with 18 oz./0.53 L capacity
    • For safe handling and convenient transportation to lab
  • Biocide-free sponge does not affect organism viability
  • Decontaminated through gamma irradiation to maintain aseptic conditions

Specifications

Product Type Dry Sponge
Dimensions (L x W) 1.5 x 3 in.
Packaging Quantity 100/Cs.
Includes Two polyethylene gloves and sample bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.