Learn More
ORS Nasco DRYBOX GLOVES L/R SZ 10.5 1PR
Supplier:  ORS Nasco 0688N3032/10H
Encompass_Preferred
Honeywell part# 8N3032/10H. Honeywell Neoprene Glove Box Gloves 8 in. Port Black Hand: Hand specific Latex Free: Yes Port Size: 20.3 cm 8 in. Size: 10.5 Thickness: 30 mil 0.76 mm Certifications/Compliance: EN 388:2016 EN 374-3 Circumference Hand: 10.5 in. 26.67 cm Diameter Cuff: 8 in. 20.3 cm Model: 8N3032/10H
Catalog No. 50-255-1654