Learn More
BUCHI Drying Rack Manifold 24
Connect up to 24 samples to a BUCHI Lyovapor™. Has 24 manifold valves and a base plate.
Supplier: BUCHI 11DCIN02000
Description
- Manifolds: adapters for traditional freeze drying beakers or round bottom flasks
- Shelves: for vials, plates, and irregular shaped items
- Heated Shelves: advanced control of drying process and applications
- Stoppering Chambers: seal vials without exposure to ambient moisture
Specifications
Benchtop Freeze Dryer | |
Ambient to -105°C | |
North America |
0 | |
Stainless Steel | |
220/240 V |