Learn More
Chemglass Life Sciences Drying Tube, Schwartz, 130mm Wide x 145mm Height
SDP
Catalog No. CG129701
Encompass_Preferred
Drying Tube, Schwartz, 130mm Wide x 145mm Height. Schwartz type drying tube having 14/20 hollow stoppers. Stoppers have a hole that aligns with a serrated hose connection for on/off action. Overall height is approximately 170mm.
Catalog No. CG129701 Supplier Chemglass Life Sciences Supplier No. CG129701
Drying Tube, Schwartz, 130mm Wide x 145mm Height. Schwartz type drying tube having 14/20 hollow stoppers. Stoppers have a hole that aligns with a serrated hose connection for on/off action. Overall height is approximately 170mm.