Learn More
StatLab Dry Mats™ Formalin Neutralizing Pads, 200 mats/case
SDP
Supplier:  StatLab 172854
Encompass_Preferred
Dry Mats™ are a unique solution for aggressive spill response and formalin vapor control. Highly absorbent and dry 12" x 13" sheets contain a proprietary formaldehyde-specific neutralizer. Excellent for grossing stations, countertops, storage cabinets, and any area where formaldehyde spills might occur.
Catalog No. NC1335470
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.