Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ DryRigger Sub-Zero Oil and Water Resistant Gloves

Catalog No. 19400089 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-400-089 X-Large
19-400-088 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-400-089 Supplier Impacto™ Supplier No. WGWINRIGGXL
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gloves repel oil and water, with 100 grams of Thinsulate™ lining for protection in cold environments. TPR padding helps protect against hairline fractures, bruising, and pinched fingers.

  • Thinsulate™ lining provides thermal protection in cold environments
  • Heavy duty TPR pad across knuckles, back of fingers, thumb & back of the hand protects against bruises & bumps
  • PVC nitrile layer offers an additional barrier of protection in oily conditions
  • Silicone dots on the palm and fingertips enhance grip capabilities
  • Ultra suede fabric in the palm repels oil & water
  • Durable patches in the webbing area assure long wear

Specifications

Back Material TPR Padding
Cuff Material Neoprene
Coating Material Silicone
Color Yellow, Black, Gray
Hand Specific
Material Suede
Product Type Anti-impact Gloves
Size X-Large
For Use With (Application) Cold Environments, Mining, Oil/Gas Extraction, and Well Drilling
Glove Cuff Color Black
Liner Material Thinsulate™
Palm Type Coated
Series DryRigger Sub-Zero
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.