Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dryzyme™ Restriction Enzymes
SDPSCI_ED

Catalog No. S94628 Shop All EDVOTEK Products
Change view
Click to view available options
Cut Site:
G.GATCC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cut Site
S94628 G.GATCC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94628 Supplier Edvotek Supplier No. 717
Add to Cart
Add to Cart

Three most frequently used restriction enzymes for biotechnology.

  • Each enzyme catalyzes cleavage at or near the defined base sequence
  • Utilized in cloning procedures
  • Restriction endonucleases are purified
  • Each lot produced meets quality control specifications

Eco RI is isolated from E. coli RY13 carrying RI (fi+ plasmid) genes for restriction and modification as well as genes for resistance to the drugs sulfathiazole and streptomycin. The recognition specificity of Eco RI is sensitive to ionic environment.

  • 5'-GAATTC-3', breaks between GA
  • 3'-CTTAAG-5', breaks between AG
Hind III - The first type restriction endonuclease activity was isolated from Haemophilus influenzae Rd cells by H. O. Smith and associates. Subsequently, the presence of two enzymes (Hind II and Hind III) in this cell strain was established.
  • 5'-AAGCTT-3', breaks between AA
  • 3'-TTCGAA-5', breaks between AA

Bam HI is isolated from Bacillus amyloliquefaciens H cells. Under certain conditions, the enzyme will recognize variations of the palindrome sequence to include GGNNCC.

  • 5'-GGATCC-3', breaks between GG
  • 3'-CCTAGG-5', breaks between GG

One restriction enzyme unit is defined as the amount of enzyme required to digest 1.0μg of lambda phage DNA in 60 min. at 37°C in a total reaction mixture of 50μL.

Order Info

  • Products are lyophilized and contain 1500 units
  • All enzyme shipments contain the appropriate reaction buffer for optimal reactions

Specifications

Concentration 500 Units
Components Appropriate reaction Buffer
Incubator Temperature 37°C
For Use With (Application) Utilized in cloning procedures
Content And Storage -20°C
Methylation Sensitivity Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not Dam Methylation-Sensitive
Enzyme BamH I
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
Type IIS RE Yes
Quantity 1500 U
Cut Site G.GATCC
Product Type BamHI
Research Category Traditional Cloning
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.