Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

QuidelOrtho VITROS™ MicroSlides

Catalog No. OCD8379034
Change view
Click to view available options
Quantity:
5 x 18 Pack
5 x 50 Pack
5 x 60 Pack
Formulation:
1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking
Alkaline Phosphatase (beef liver, E.C.3.1.3.1) 0.02U, Magnesium Chloride 3μg and p-Nitrophenyl Phosphate 0.3mg, Pigment, Binders, Surfactants, Buffer, Enzyme cofactor, protein, Stabilizer and cross-linking agent
Copper Sulfate 0.9mg, Tartaric Acid 1.2mg and Lithium Hydroxide 1.3mg, Polymer beads, Binders and Surfactants
Not Applicable
Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.2mg, Potassium Chloride 63μg and Valinomycin 55μg, Binders, Plasticizers, Stabilizer, Surfactants and Nickel-chromium
Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.3mg, Methyl Monensin 50μg, Binders, Buffer, Plasticizers, Stabilizer, Surfactants and Nickel-chromium
Uricase (Candida utilis, E.C.1.7.3.3) 0.02U, Peroxidase (horseradish root, E.C.1.11.1.7) 0.6U and 2-(3,5-Dimethoxy-4-Hydroxyphenyl)-4,5-bis-(4-Dimethylaminophenyl) Imidazole (leucodye) 14μg, Stabilizers, Pigment, Binders, Buffer, Surfact
p-Methylaminophenol Sulfate 350μg, Ammonium Molybdate 340μg, Pigment, Binders, Surfactants, Buffer and Stabilizers
Type:
18-Magnesium Dry Slide
18-Theophylline Dry Slide
Direct HDL Cholesterol-dHDL
Magnesium Dry Slide
Phosphorus Dry Slide
Potassium Dry Slide
Sodium Dry Slide
Total Protein Dry Slide
Uric Acid Dry Slide
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation Type
OCD8379034 5 x 50 Pack Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.3mg, Methyl Monensin 50μg, Binders, Buffer, Plasticizers, Stabilizer, Surfactants and Nickel-chromium Sodium Dry Slide
OCD1921204 5 x 18 Pack 1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking 18-Magnesium Dry Slide
OCD8157596 5 x 50 Pack Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.2mg, Potassium Chloride 63μg and Valinomycin 55μg, Binders, Plasticizers, Stabilizer, Surfactants and Nickel-chromium Potassium Dry Slide
OCD1513209 5 x 60 Pack p-Methylaminophenol Sulfate 350μg, Ammonium Molybdate 340μg, Pigment, Binders, Surfactants, Buffer and Stabilizers Phosphorus Dry Slide
OCD1943927 5 x 60 Pack Uricase (Candida utilis, E.C.1.7.3.3) 0.02U, Peroxidase (horseradish root, E.C.1.11.1.7) 0.6U and 2-(3,5-Dimethoxy-4-Hydroxyphenyl)-4,5-bis-(4-Dimethylaminophenyl) Imidazole (leucodye) 14μg, Stabilizers, Pigment, Binders, Buffer, Surfact Uric Acid Dry Slide
OCD8392292 5 x 50 Pack Copper Sulfate 0.9mg, Tartaric Acid 1.2mg and Lithium Hydroxide 1.3mg, Polymer beads, Binders and Surfactants Total Protein Dry Slide
OCD8255093 5 x 60 Pack 1,2-bis(o-Aminophenoxy)ethane-N,N,N',N'-Tetraacetic Acid (Calcium chelator) 242μg, 1,5-bis(2-Hydroxy-3,5-Dichlorophenyl)-3-Cyanoformazan (dye) 38μg, Pigment, Binders, Buffer, dye solubilizer, Surfactants, cross-linking Magnesium Dry Slide
OCD1307164 5 x 18 Pack Alkaline Phosphatase (beef liver, E.C.3.1.3.1) 0.02U, Magnesium Chloride 3μg and p-Nitrophenyl Phosphate 0.3mg, Pigment, Binders, Surfactants, Buffer, Enzyme cofactor, protein, Stabilizer and cross-linking agent 18-Theophylline Dry Slide
OCD6802469 5 x 18 Pack Not Applicable Direct HDL Cholesterol-dHDL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. OCD8379034 Supplier QuidelOrtho Supplier No. 8379034
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with the QuidelOrtho VITROS™ 250 System

Specifications

Sample Type Heparin, Plasma, Serum, Urine
Type Sodium Dry Slide
Formulation Silver 0.4mg, Silver Chloride 0.2mg, Sodium Chloride 0.3mg, Methyl Monensin 50μg, Binders, Buffer, Plasticizers, Stabilizer, Surfactants and Nickel-chromium
For Use With (Application) In-vitro diagnostic use
Quantity 5 x 50 Pack
Sample Size 10 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.