Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DsbA Monoclonal Antibody (8A10)
GREENER_CHOICE

Catalog No. pima518312
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518312 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518312 Supplier Invitrogen™ Supplier No. MA518312
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody detects endogenous levels of DsbA at a molecular weight of 24 kDa and does not cross-react with related proteins. Purity is >95% by SDS-PAGE.

DsbA is required for disulfide bond formation in some periplasmic proteins such as PhoA or OmpA. Acts by transferring its disulfide bond to other proteins and is reduced in the process. DsbA is reoxidized by DsbB. It is required for pilus biogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DsbA
Applications Western Blot
Classification Monoclonal
Clone 8A10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Alias Disulphide oxidoreductase; Thiol disulfide interchange protein
Host Species Mouse
Immunogen Recombinant full length DsbA from E. coli.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.