Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DSE Mouse anti-Human, Mouse, Clone: 2C9F7, Proteintech
SDP

Catalog No. 501736200 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
Catalog No. Quantity
50-173-6200 150 μL
1 options

Catalog No. 50-173-6200

Supplier: Proteintech Group Inc 600411IG150UL

Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Converts D-glucuronic acid to L-iduronic acid (IdoUA) residues.

Specifications

Antigen DSE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2C9F7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DSE
Gene Accession No. Q8BLI4, Q9UL01
Gene Alias dermatan sulfate epimerase, DS epimerase, DSE, DSEPI, SART 2, SART2
Gene Symbols Dse
Host Species Mouse
Immunogen DSE Fusion Protein Ag0695
Purification Method Caprylic Acid Precipitation
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 212898, 29940
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.