Learn More
Creative Pegworks DSPE-PEG-COOH, MW 3.4K 50MG
SDP
Supplier:  Creative Pegworks PLS991350MG
Encompass_Preferred
DSPE-PEG-COOH is a linear heterobifunctional PEGylation reagent with a DSPE phospholipid and a carboxylic acid. It is a useful self-assembling reagent to prepare PEGylated liposome or micelle while also providing a COOH group for conjugation with amine-containing molecules.50mg CREATIVE PEGWORKS mfg#PLS-9913
Catalog No. NC1962498
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.