Learn More
Electron Microscopy Sciences Disposable Scalpel Handle Set #20
SDP
Supplier:  Electron Microscopy Sciences 7205220
Encompass_Preferred
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation. They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Catalog No. 50-190-0391
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.