Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynex™ DSX™ Automated ELISA Processor

Catalog No. 23333002
Change view
Click to view available options
Description:
2 plates system
4 plates system
4 plates system (with no incubator)
Incubator Module
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-333-002 4 plates system (with no incubator)
23-333-003 2 plates system
23-333-004 4 plates system
23-333-005 Incubator Module
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Appropriate precautions must be taken when working with biohazardous material.
Catalog No. 23-333-002 Supplier Dynex™ Supplier No. 65100
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy
Appropriate precautions must be taken when working with biohazardous material.

Busy clinical and medium-throughput private and industrial labs require advanced automation that can handle virtually every ELISA immunoassay, all in a reliable, cost-effective package. Look no further than the DSX™

  • Completely automated open system
  • Well fill verification
  • Unique dispense synchronization
  • 96 on-board samples
  • Up to 12 different assays simultaneously
  • Automatic data reduction
  • Easy-to-Use Revelation DSX Software
  • Disposable sample and reagent tips
  • Specifications

    Type Open System
    Description 4 plates system (with no incubator)
    For Use With (Application) ELISA Assays
    Sample Dilution (192) 24 x 8 deep-well strips Maximum dilution: 1 to 36,100
    Throughput Assay dependent
    Analytical Method Photometric
    Capacity 98 samples on-board
    Certifications/Compliance EN 61326-1:2013EN 61326-2-6:2013 IEC/EN 61010-1:2010 Ed.3 IEC/EN 61010-2-010:2014 Ed.3 IEC/EN 61010-2-101 issued 2015/08/14 Ed. 2 CSA C22.2 No. 61010-1:2012 Ed.3 CSA C22.2 No. 61010-2-101:2015 Ed.2 RoHS Compliant
    Includes One year warranty against defects in parts, materials, and workmanship
    Wavelength 405/690 nm
    Depth (English) 36 in.
    Depth (Metric) 91 cm
    Height (English) 32 in.
    Height (Metric) 81 cm
    Width (English) 42 in.
    Width (Metric) 106 cm
    Electrical Requirements 100/240 V auto-switching
    Frequency 50/60 Hz
    Data Management Features Revelation DSX™ Software
    Operating System Windows 7/ Windows 10
    Measurement Method Absorbance
    Weight (English) 537 lb.
    Weight (Metric) 244 kg
    Show More Show Less

    DYNEX makes no representations, warranties or performance claims with respect to the performance of Agility for any specific application, including clinical application or for the use of the Agility system with any reagents, assays or other products.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.