Learn More
Dynex™ DSX™/ DS2™/ Agility™ Accessories
Catalog No. 23333015
Change view
Click to view available options
Description:
24 Reagent Bottles
450 nm filter
630 nm filter
Deep Well Dilution Strips
Inkjet Printer
SU1000XLNES Smart UPS
Standard Control Bottles
Verification Plate
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
23-333-015 Verification Plate
23-710-000 Deep Well Dilution Strips
23-710-007 24 Reagent Bottles
23-710-006 Standard Control Bottles
23-710-021 450 nm filter
23-710-020 630 nm filter
23-333-013 SU1000XLNES Smart UPS
23-333-011 Inkjet Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 23-333-015 Supplier Dynex™ Supplier No. 24099
Add to Cart
Add to Cart
Dynex™ certified accessories for use with DSX™, DS2™, or Agility™
  • Dynex Certified Consumables for use with DSX™, DS2™, or Agility™
  • Accessories used are integral components of the controlled system
  • Only Dynex Certified Consumables are specifically designed and produced for Dynex instruments, ensuring proper tip fit with superior accuracy and performance
  • Busy clinical and medium-throughput private and industrial labs require advanced automation that can handle virtually every ELISA immunoassay, all in a reliable, cost-effective package; look no further than the DSX™
  • Automation, accuracy, immediacy and responsiveness are the must-haves of demanding near-patient-care settings and lower-throughput labs. Finding all this in a reliable, cost-effective and easy to use and maintain package is possible with the DS2™
  • Agility's™ intelligent design represents the next breakthrough in ELISA automation
  • Specifications
    Description Verification Plate
    Includes alibration at 405, 450, 490, 550, 620, 630 nm